Gene Transcripts

Gene Symbol
ZDHHC9
Entrez Gene ID
51114
Gene Name
zinc finger DHHC-type containing 9
Gene Aliases
CGI89, CXorf11, DHHC9, MMSA1, MRXSZ, ZDHHC10, ZNF379, ZNF380
Chromosome Location
Chr. X: 129803288 - 129843934 on Build GRCh38
UniGene ID
Hs.193566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008222.2 NP_001008223.1 2 529
NM_016032.3 NP_057116.2 3 509
XM_011531348.2 XP_011529650.1 3 496
GenBank mRNA AB209807.1 - 2 448
AF151847.1 - 3 397
AK001112.1 - 3 459
AK001524.1 - 2 260
AL161962.1 - 3 386
AY358558.1 - 3 441
AY952881.8 - 3 481
BC000035.2 - 2 403 3506601
BC003128.1 - 3 447 2957911
BC006200.2 - 3 443 2964425
BC012826.2 - 3 443 2964425
BM909699.1 - 436
CD627424.1 - 3 474
DA493628.1 - 3 509
DA768650.1 - 2 529

View Full Product Details