Gene Transcripts

Gene Symbol
RNF8
Entrez Gene ID
9025
Gene Name
ring finger protein 8
Gene Aliases
hRNF8
Chromosome Location
Chr. 6: 37353972 - 37394738 on Build GRCh38
UniGene ID
Hs.485278
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003958.3 NP_003949.1 8 4254
NM_183078.2 NP_898901.1 7 4049
NR_046399.1 8 4360
GenBank mRNA AB014546.1 - 8 4173

View Full Product Details