Gene Transcripts

Gene Symbol
NUBP2
Entrez Gene ID
10101
Gene Name
nucleotide binding protein 2
Gene Aliases
CFD1, NBP 2, NUBP1
Chromosome Location
Chr. 16: 1782923 - 1789191 on Build GRCh38
UniGene ID
Hs.256549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF118394.1 - 7 1295
AK001023.1 - 7 1305
AK093593.1 - 5 2142
BC002768.2 - 7 1286 3633393
BC008005.1 - 7 1311 3507768
BX648907.1 - 5 2097

View Full Product Details