Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143850.2 | NP_001137322.1 | 10, 11 | 1367 | |
NM_024809.4 | NP_079085.2 | 10, 11 | 1370 | ||
XM_006719605.3 | XP_006719668.1 | 10, 11 | 1370 | ||
XM_017019975.1 | XP_016875464.1 | 9, 10 | 1245 | ||
GenBank mRNA | AK056924.1 | - | 10, 11 | 1368 | |
AK292153.1 | - | 10, 11 | 1367 | ||
BC009112.1 | - | 10, 11 | 1339 | 3463475 | |
DQ278870.1 | - | 10, 11 | 1368 |