Gene Transcripts

Gene Symbol
APOPT1
Entrez Gene ID
84334
Gene Name
apoptogenic 1, mitochondrial
Gene Aliases
APOP, APOP1, C14orf153
Chromosome Location
Chr. 14: 103562957 - 103592187 on Build GRCh38
UniGene ID
Hs.598441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032374.4 NP_115750.2 5 1527
GenBank mRNA BC007412.2 - 5 1490

View Full Product Details