Gene Transcripts

Gene Symbol
Trim21
Entrez Gene ID
20821
Gene Name
tripartite motif-containing 21
Gene Aliases
Ro52, Ssa1
Chromosome Location
Chr. 7: 102557920 - 102565482 on Build GRCm38
UniGene ID
Mm.321227
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082552.2 NP_001076021.1 4 830
NM_009277.4 NP_033303.3 5 899
GenBank mRNA AK141294.1 - 5 879
AK150484.1 - 4 794
AK152619.1 - 4 794
AK171489.1 - 5 864
BC010580.1 - 5 839 3584654
CT010336.1 - 3 724
L27990.1 - 5 864

View Full Product Details