Gene Transcripts

Gene Symbol
Bnc2
Entrez Gene ID
242509
Gene Name
basonuclin 2
Gene Aliases
5031434M05Rik, 8430420F16Rik
Chromosome Location
Chr. 4: 84266804 - 84676342 on Build GRCm38
UniGene ID
Mm.190774
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK031440.1 - 4 423
AY438375.1 - 1 113
BC110483.2 - 1 102 40048720
BC110484.1 - 1 102 40048722

View Full Product Details