Gene Transcripts
- Gene Symbol
- Atp9a
- Entrez Gene ID
- 11981
- Gene Name
- ATPase, class II, type 9A
- Gene Aliases
- IIa
- Chromosome Location
- Chr. 2: 168634438 - 168742136 on Build GRCm38
- UniGene ID
- Mm.330819
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289445.1 |
NP_001276374.1
|
20, 21 |
2464 |
|
NM_001289446.1 |
NP_001276375.1
|
20, 21 |
2194 |
|
NM_015731.3 |
NP_056546.2
|
20, 21 |
2264 |
|
GenBank mRNA |
AF152243.1 |
- |
20, 21 |
2432 |
|
AK082271.1 |
- |
20, 21 |
2190 |
|
AK089064.1 |
- |
20, 21 |
2370 |
|
AK177735.1 |
- |
1, 2 |
89 |
|
AK204681.1 |
- |
2, 3 |
160 |
|
BC006949.1 |
- |
18, 19 |
1966 |
|
BC075718.1 |
- |
20, 21 |
2392 |
|
BC094551.1 |
- |
20, 21 |
2234 |
6813369 |
View Full Product Details