Gene Transcripts

Gene Symbol
Arhgef12
Entrez Gene ID
69632
Gene Name
Rho guanine nucleotide exchange factor (GEF) 12
Gene Aliases
2310014B11Rik, AU019857, Larg, mKIAA0382
Chromosome Location
Chr. 9: 42963842 - 43105718 on Build GRCm38
UniGene ID
Mm.275266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027144.2 NP_081420.2 3 628
GenBank mRNA AF467766.1 - 3 393
AK009331.1 - 3 602
AK122267.1 - 3 628
BC052785.1 - 173

View Full Product Details