Gene Transcripts
- Gene Symbol
- Negr1
- Entrez Gene ID
- 320840
- Gene Name
- neuronal growth regulator 1
- Gene Aliases
- 5330422G01Rik, KILON, Ntra
- Chromosome Location
- Chr. 3: 156561794 - 157316464 on Build GRCm38
- UniGene ID
- Mm.317293
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039094.3 |
NP_001034183.1
|
3, 4 |
926 |
|
NM_177274.4 |
NP_796248.1
|
3, 4 |
926 |
|
GenBank mRNA |
AJ487032.1 |
- |
3, 4 |
546 |
|
AK030503.1 |
- |
3, 4 |
752 |
|
AK035218.1 |
- |
3, 4 |
762 |
|
AK122576.2 |
- |
2, 3 |
269 |
|
AK142298.1 |
- |
3, 4 |
620 |
|
AK147253.1 |
- |
3, 4 |
927 |
|
BC146005.1 |
- |
3, 4 |
711 |
40131308 |
BC146007.1 |
- |
3, 4 |
710 |
40131310 |
View Full Product Details