Gene Transcripts

Gene Symbol
Ankrd61
Entrez Gene ID
66729
Gene Name
ankyrin repeat domain 61
Gene Aliases
4921520G13Rik
Chromosome Location
Chr. 5: 143890741 - 143895152 on Build GRCm38
UniGene ID
Mm.439817
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025732.3 NP_080008.1 1 282
GenBank mRNA AK014929.1 - 1 283
AK017053.1 - 1 223
BC052346.1 - 1 245 6703467

View Full Product Details