Gene Transcripts

Gene Symbol
Abt1
Entrez Gene ID
30946
Gene Name
activator of basal transcription 1
Gene Aliases
Chromosome Location
Chr. 13: 23418361 - 23423866 on Build GRCm38
UniGene ID
Mm.292094
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013924.3 NP_038952.1 2 346
GenBank mRNA AB021860.1 - 2 345
AK009557.1 - 2 347
AK029222.1 - 2 347
BC082609.1 - 2 339 30680713

View Full Product Details