Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253381.1 | NP_001240310.1 | 8 | 1995 | |
NM_017377.5 | NP_059073.1 | 8 | 1960 | ||
GenBank mRNA | AF142670.1 | - | 7 | 1603 | |
AK019111.1 | - | 1 | 116 | ||
AK145250.1 | - | 9 | 2017 | ||
BC138567.1 | - | 7 | 1596 | 8861587 | |
BC138569.1 | - | 7 | 1596 | 8861589 | |
BC145335.1 | - | 7 | 1631 | 9053891 | |
BX526030.1 | - | 1 | 447 |