Gene Transcripts
- Gene Symbol
- Ppp1r9b
- Entrez Gene ID
- 84686
- Gene Name
- protein phosphatase 1, regulatory subunit 9B
- Gene Aliases
- Neb2
- Chromosome Location
- Chr. 10: 82800704 - 82816735 on Build Rnor_6.0
- UniGene ID
- Rn.6764
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053474.1 |
NP_445926.1
|
11 |
3936 |
|
GenBank mRNA |
AF016252.1 |
- |
11 |
3936 |
|
View Full Product Details