Gene Transcripts

Gene Symbol
Ccdc50
Entrez Gene ID
288022
Gene Name
coiled-coil domain containing 50
Gene Aliases
C3orf6h, c3orf6
Chromosome Location
Chr. 11: 76742179 - 76804698 on Build Rnor_6.0
UniGene ID
Rn.14503
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182736.1 NP_877400.1 8 666
XM_008768795.1 XP_008767017.1 9 1431
GenBank mRNA AJ534984.1 - 8 666

View Full Product Details