Gene Transcripts

Gene Symbol
Spata20
Entrez Gene ID
360604
Gene Name
spermatogenesis associated 20
Gene Aliases
Ssp411
Chromosome Location
Chr. 10: 82202619 - 82217259 on Build Rnor_6.0
UniGene ID
Rn.50006
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199402.1 NP_955434.1 16 2559
XM_006247200.2 XP_006247262.1 16 2585
GenBank mRNA AY438568.1 - 16 2559

View Full Product Details