Gene Transcripts

Gene Symbol
OR51G1
Entrez Gene ID
79324
Gene Name
olfactory receptor family 51 subfamily G member 1 (gene/pseudogene)
Gene Aliases
OR11-29, OR51G3P
Chromosome Location
Chr. 11: 4923374 - 4924339 on Build GRCh38
UniGene ID
Hs.554541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005237.1 NP_001005237.1 1 325
GenBank mRNA BC136852.1 - 1 393
BC136861.1 - 1 393 9020852

View Full Product Details