Gene Transcripts

Gene Symbol
Vom1r38
Entrez Gene ID
494307
Gene Name
vomeronasal 1 receptor 38
Gene Aliases
V1rd25
Chromosome Location
Chr. 1: 72230038 - 72230961 on Build Rnor_6.0
UniGene ID
Rn.129427
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008964.1 NP_001008964.1 1 571
GenBank mRNA AY510334.1 - 1 571

View Full Product Details