Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015439.2 | NP_056254.1 | 2 | 456 | |
XM_011535728.2 | XP_011534030.1 | 2 | 459 | ||
GenBank mRNA | AJ420568.1 | - | 2 | 207 | |
AK300672.1 | - | 2 | 207 | ||
AL050197.1 | - | 2 | 206 | ||
AY167571.1 | - | 2 | 474 | ||
BC004464.1 | - | 2 | 190 | 2820737 | |
BC013019.2 | - | 2 | 188 | ||
BC080566.1 | - | 2 | 190 | ||
BC108717.1 | - | 2 | 379 | 6068596 |