Gene Transcripts
- Gene Symbol
- CCDC53
- Entrez Gene ID
- 51019
- Gene Name
- coiled-coil domain containing 53
- Gene Aliases
- CGI-116
- Chromosome Location
- Chr. 12: 102012840 - 102062124 on Build GRCh38
- UniGene ID
- Hs.405692
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301107.1 |
NP_001288036.1
|
2, 3 |
317 |
|
NM_016053.3 |
NP_057137.1
|
2, 3 |
317 |
|
XM_011538423.1 |
XP_011536725.1
|
2, 3 |
235 |
|
GenBank mRNA |
AF151874.1 |
- |
2, 3 |
262 |
|
AF155655.1 |
- |
2, 3 |
262 |
|
AK223339.1 |
- |
2, 3 |
253 |
|
AK314970.1 |
- |
2, 3 |
235 |
|
BC010889.1 |
- |
2, 3 |
188 |
4272385 |
BX356987.2 |
- |
2, 3 |
272 |
|
CR457171.1 |
- |
2, 3 |
155 |
|
DB452335.1 |
- |
2, 3 |
317 |
|
View Full Product Details