Gene Transcripts

Gene Symbol
C16orf52
Entrez Gene ID
730094
Gene Name
chromosome 16 open reading frame 52
Gene Aliases
Chromosome Location
Chr. 16: 22008135 - 22084654 on Build GRCh38
UniGene ID
Hs.498890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164579.1 NP_001158051.1 2 367
GenBank mRNA BC027604.1 - 3 397
BC053950.1 - 3 384

View Full Product Details