Gene Transcripts

Gene Symbol
Adprhl1
Entrez Gene ID
290880
Gene Name
ADP-ribosylhydrolase like 1
Gene Aliases
Chromosome Location
Chr. 16: 81616130 - 81648133 on Build Rnor_6.0
UniGene ID
Rn.144754
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013054.1 NP_001013072.1 7 1259
GenBank mRNA BC083773.1 - 7 1259

View Full Product Details