Gene Transcripts
- Gene Symbol
- LINC01465
- Entrez Gene ID
- 283416
- Gene Name
- long intergenic non-protein coding RNA 1465
- Gene Aliases
- C12orf61
- Chromosome Location
- Chr. 12: 62601751 - 62603434 on Build GRCh38
- UniGene ID
- Hs.375881
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_121682.1 |
|
1 |
1391 |
|
GenBank mRNA |
AK098456.1 |
- |
1 |
1392 |
|
View Full Product Details