Gene Transcripts

Gene Symbol
Slfnl1
Entrez Gene ID
500540
Gene Name
schlafen-like 1
Gene Aliases
Chromosome Location
Chr. 5: 139466895 - 139473599 on Build Rnor_6.0
UniGene ID
Rn.47152
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024347.1 NP_001019518.1 1737
GenBank mRNA BC079280.1 - 1737

View Full Product Details