Gene Transcripts

Gene Symbol
Paqr9
Entrez Gene ID
75552
Gene Name
progestin and adipoQ receptor family member IX
Gene Aliases
1700020G04Rik
Chromosome Location
Chr. 9: 95559817 - 95562121 on Build GRCm38
UniGene ID
Mm.151485
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198414.2 NP_940806.2 1 1585
GenBank mRNA AK006159.1 - 1 133
AY424298.1 - 1 1585

View Full Product Details