Gene Transcripts

Gene Symbol
Arrdc3
Entrez Gene ID
309945
Gene Name
arrestin domain containing 3
Gene Aliases
LRRGT00048
Chromosome Location
Chr. 2: 8732910 - 8745448 on Build Rnor_6.0
UniGene ID
Rn.161886
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007797.1 NP_001007798.1 7 1128
GenBank mRNA AY383703.1 - 7 1128

View Full Product Details