Gene Transcripts

Gene Symbol
Pmaip1
Entrez Gene ID
492821
Gene Name
phorbol-12-myristate-13-acetate-induced protein 1
Gene Aliases
Noxa
Chromosome Location
Chr. 18: 62174850 - 62178837 on Build Rnor_6.0
UniGene ID
Rn.163380
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008385.1 NP_001008386.1 2, 3 224
GenBank mRNA AY788892.1 - 2, 3 224

View Full Product Details