Gene Transcripts

Gene Symbol
C17orf67
Entrez Gene ID
339210
Gene Name
chromosome 17 open reading frame 67
Gene Aliases
Chromosome Location
Chr. 17: 56791913 - 56833990 on Build GRCh38
UniGene ID
Hs.658949
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085430.2 NP_001078899.2 7 1538
GenBank mRNA BC041467.1 - 7 1538
BC093905.1 - 3 338 7939750
BC093907.1 - 3 338 7939752

View Full Product Details