Gene Transcripts

Gene Symbol
C5orf63
Entrez Gene ID
401207
Gene Name
chromosome 5 open reading frame 63
Gene Aliases
YDR286C
Chromosome Location
Chr. 5: 127040752 - 127073539 on Build GRCh38
UniGene ID
Hs.49573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164479.1 NP_001157951.1 5 989
GenBank mRNA AK126569.1 - 6 1087

View Full Product Details