Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099218.2 | NP_001092688.1 | 1 | 77 | |
NM_001321233.1 | NP_001308162.1 | 5 | 435 | ||
XM_011533084.1 | XP_011531386.1 | 1 | 77 | ||
GenBank mRNA | AK310498.1 | - | 6 | 682 | |
DQ860102.1 | - | 1 | 77 |