Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256169.1 | NP_001243098.1 | 2 | 210 | |
NM_019101.2 | NP_061974.2 | 2 | 255 | ||
NR_045828.1 | 2 | 210 | |||
XM_006715150.3 | XP_006715213.1 | 2 | 232 | ||
GenBank mRNA | AF118393.3 | - | 2 | 215 | |
AF161454.1 | - | 2 | 176 | ||
AJ245434.1 | - | 2 | 231 | ||
BC020683.1 | - | 2 | 234 | 4733993 | |
BG564719.1 | - | 2 | 256 | ||
CB054016.1 | - | 2 | 217 | ||
CN428415.1 | - | 2 | 197 |