Gene Transcripts

Gene Symbol
Ankrd16
Entrez Gene ID
307102
Gene Name
ankyrin repeat domain 16
Gene Aliases
Chromosome Location
Chr. 17: 70384883 - 70398089 on Build Rnor_6.0
UniGene ID
Rn.45470
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033698.1 NP_001028870.1 4 680
GenBank mRNA BC099837.1 - 4 680

View Full Product Details