Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130156.2 | NP_001123628.1 | 5 | 735 | |
NM_001130157.2 | NP_001123629.1 | 6 | 808 | ||
NM_001195432.1 | NP_001182361.1 | 7 | 1010 | ||
NM_001195433.1 | NP_001182362.1 | 5 | 679 | ||
NM_001195434.1 | NP_001182363.1 | 7 | 853 | ||
NM_022443.4 | NP_071888.1 | 5 | 691 | ||
XM_005247479.4 | XP_005247536.1 | 6 | 738 | ||
XM_005247480.2 | XP_005247537.1 | 6 | 722 | ||
XM_006713645.2 | XP_006713708.1 | 4 | 529 | ||
XM_011512852.2 | XP_011511154.1 | 7 | 811 | ||
XM_011512853.2 | XP_011511155.1 | 6 | 758 | ||
XM_011512854.2 | XP_011511156.1 | 4 | 527 | ||
GenBank mRNA | AK056948.1 | - | 7 | 823 | |
AK096889.1 | - | 7 | 919 | ||
AK297488.1 | - | 5 | 677 | ||
AK315402.1 | - | 5 | 661 | ||
AY848700.1 | - | 5 | 648 | ||
AY848701.1 | - | 6 | 777 | ||
AY848702.1 | - | 5 | 704 | ||
BC007045.1 | - | 5 | 660 | 3996145 | |
BX647666.1 | - | 6 | 746 | ||
KU178186.1 | - | 5 | 554 | ||
KU178187.1 | - | 6 | 599 | ||
KU178188.1 | - | 4 | 406 | ||
L49054.1 | - | 5 | 662 |