Gene Transcripts

Gene Symbol
AGMAT
Entrez Gene ID
79814
Gene Name
agmatinase
Gene Aliases
Chromosome Location
Chr. 1: 15571699 - 15585110 on Build GRCh38
UniGene ID
Hs.461532
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024758.4 NP_079034.3 5 953
GenBank mRNA AK130034.1 - 5 749
AK225366.1 - 5 866
AY057097.1 - 5 853
BC005090.1 - 5 953 3837251

View Full Product Details