Gene Transcripts
- Gene Symbol
- ST6GAL2
- Entrez Gene ID
- 84620
- Gene Name
- ST6 beta-galactoside alpha-2,6-sialyltransferase 2
- Gene Aliases
- SIAT2, ST6GalII
- Chromosome Location
- Chr. 2: 106801600 - 106887410 on Build GRCh38
- UniGene ID
- Hs.98265
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142351.1 |
NP_001135823.1
|
2, 3 |
1063 |
|
NM_001142352.1 |
NP_001135824.1
|
2, 3 |
1133 |
|
NM_001322362.1 |
NP_001309291.1
|
2, 3 |
1112 |
|
NM_032528.2 |
NP_115917.1
|
2, 3 |
1133 |
|
XM_006712802.1 |
XP_006712865.1
|
2, 3 |
1148 |
|
XM_011511999.2 |
XP_011510301.1
|
2, 3 |
1245 |
|
XM_011512000.2 |
XP_011510302.1
|
2, 3 |
1330 |
|
XM_011512001.2 |
XP_011510303.1
|
2, 3 |
1330 |
|
XM_017005110.1 |
XP_016860599.1
|
2, 3 |
1084 |
|
GenBank mRNA |
AB058780.1 |
- |
2, 3 |
1063 |
|
AB059555.1 |
- |
2, 3 |
1119 |
|
AK307784.1 |
- |
2, 3 |
1133 |
|
BC008680.1 |
- |
2, 3 |
1119 |
3856171 |
View Full Product Details