Gene Transcripts

Gene Symbol
NFATC1
Entrez Gene ID
4772
Gene Name
nuclear factor of activated T-cells 1
Gene Aliases
NF-ATC, NF-ATc1.2, NFAT2, NFATc
Chromosome Location
Chr. 18: 79395772 - 79529323 on Build GRCh38
UniGene ID
Hs.534074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278669.1 NP_001265598.1 6 2339
NM_001278670.1 NP_001265599.1 6 2339
NM_001278672.1 NP_001265601.1 6 1965
NM_001278673.1 NP_001265602.1 5 866
NM_001278675.1 NP_001265604.1 6 1965
NM_006162.4 NP_006153.2 6 2339
NM_172387.2 NP_765975.1 6 1965
NM_172388.2 NP_765976.1 5 1240
NM_172389.2 NP_765977.1 6 1965
NM_172390.2 NP_765978.1 6 2339
XM_017025783.1 XP_016881272.1 6 2181
GenBank mRNA AK292641.1 - 6 2181
AK293760.1 - 5 811
BC104753.1 - 6 1980 8143756
BC112243.1 - 6 1980 8327711
BC143457.1 - 6 1895
EU887559.1 - 6 1886
EU887560.1 - 6 1847
EU887561.1 - 6 1886
EU887562.1 - 6 1847
EU887563.1 - 6 1886
EU887564.1 - 6 1847
EU887565.1 - 6 1886
EU887566.1 - 6 1847
U08015.1 - 6 2125
U59736.1 - 6 1922
U80917.1 - 6 2255
U80918.1 - 6 1909
U80919.1 - 5 1068

View Full Product Details