Gene Transcripts
- Gene Symbol
- C16orf52
- Entrez Gene ID
- 730094
- Gene Name
- chromosome 16 open reading frame 52
- Gene Aliases
- Chromosome Location
- Chr. 16: 22008135 - 22084654 on Build GRCh38
- UniGene ID
- Hs.498890
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC027604.1 |
- |
5, 6 |
773 |
|
BC053950.1 |
- |
5, 6 |
760 |
|
View Full Product Details