Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001303252.1 | NP_001290181.1 | 4 | 651 | |
NM_002969.4 | NP_002960.2 | 4 | 651 | ||
GenBank mRNA | AK307421.1 | - | 3 | 239 | |
BC015741.1 | - | 4 | 552 | 4907996 | |
BX640648.1 | - | 3 | 288 | ||
CB123561.1 | - | 3 | 224 | ||
CR456515.1 | - | 4 | 436 | ||
U66243.1 | - | 4 | 803 | ||
X79483.1 | - | 4 | 434 | ||
Y10487.1 | - | 4 | 607 |