Gene Transcripts

Gene Symbol
NME7
Entrez Gene ID
29922
Gene Name
NME/NM23 family member 7
Gene Aliases
CFAP67, MN23H7, NDK 7, NDK7, nm23-H7
Chromosome Location
Chr. 1: 169132385 - 169367963 on Build GRCh38
UniGene ID
Hs.706952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013330.4 NP_037462.1 3 384
NM_197972.2 NP_932076.1 3 353
NR_104229.1 3 384
XM_017001099.1 XP_016856588.1 3 370
GenBank mRNA AB209049.1 - 3 193
AF153191.2 - 3 223
AK094513.1 - 3 336
AK290701.1 - 3 218
AK301917.1 - 3 367
BC006983.1 - 3 215 3997152
CD107958.1 - 3 412

View Full Product Details