Gene Transcripts

Gene Symbol
Vom2r40
Entrez Gene ID
286983
Gene Name
vomeronasal 2 receptor, 40
Gene Aliases
Chromosome Location
Chr. 1: 148927532 - 148974509 on Build Rnor_6.0
UniGene ID
Rn.10808
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173317.1 NP_775439.1 1288
GenBank mRNA AF016180.1 - 1288

Gene Symbol
LOC679574
Entrez Gene ID
679574
Gene Name
similar to putative pheromone receptor (Go-VN3)
Gene Aliases
Chromosome Location
Chr. 1: 149437496 - 149476747 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759645.1 XP_008757867.1 3 648

Gene Symbol
Vom2r42
Entrez Gene ID
679557
Gene Name
vomeronasal 2 receptor, 42
Gene Aliases
Chromosome Location
Chr. 1: 149132320 - 149175214 on Build Rnor_6.0
UniGene ID
Rn.217635
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099511.1 NP_001092981.1 3 666

View Full Product Details