Gene Transcripts

Gene Symbol
Nubpl
Entrez Gene ID
76826
Gene Name
nucleotide binding protein-like
Gene Aliases
2410170E07Rik
Chromosome Location
Chr. 12: 52097746 - 52310970 on Build GRCm38
UniGene ID
Mm.244781
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029760.2 NP_084036.2 10 898
GenBank mRNA AK010827.1 - 10 880
AK141257.1 - 10 862
AK152377.1 - 10 899
BC023294.1 - 492
BC062175.1 - 10 879 30291107

View Full Product Details