Gene Transcripts
- Gene Symbol
- Timm17a
- Entrez Gene ID
- 54311
- Gene Name
- translocase of inner mitochondrial membrane 17 homolog A (yeast)
- Gene Aliases
- Tim17, Timm17
- Chromosome Location
- Chr. 13: 52124641 - 52136127 on Build Rnor_6.0
- UniGene ID
- Rn.22514
- Species
- Rattus norvegicus
- Gene Symbol
- Timm17al1
- Entrez Gene ID
- 500655
- Gene Name
- translocase of inner mitochondrial membrane 17a-like 1
- Gene Aliases
- RGD1560665
- Chromosome Location
- Chr. 6: 83252019 - 83253099 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_576033.5 |
XP_576033.1
|
2 |
127 |
|
View Full Product Details