Gene Transcripts

Gene Symbol
Apom
Entrez Gene ID
55939
Gene Name
apolipoprotein M
Gene Aliases
Chromosome Location
Chr. 20: 5120473 - 5123073 on Build Rnor_6.0
UniGene ID
Rn.262
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019373.2 NP_062246.1 2 281
GenBank mRNA AF207821.1 - 2 258
BC089807.1 - 2 230 7323885
FQ210093.1 - 2 280
FQ219449.1 - 2 281

View Full Product Details