Gene Transcripts

Gene Symbol
CRCP
Entrez Gene ID
27297
Gene Name
CGRP receptor component
Gene Aliases
CGRP-RCP, CGRPRCP, RCP, RCP9
Chromosome Location
Chr. 7: 66114818 - 66154568 on Build GRCh38
UniGene ID
Hs.300684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040647.1 NP_001035737.1 5 384
NM_001040648.1 NP_001035738.1 3 252
NM_001142414.1 NP_001135886.1 5 328
NM_014478.4 NP_055293.1 6 483
NR_024548.1 7 605
GenBank mRNA AF073792.1 - 5 338
AK290553.1 - 5 352
AK303737.1 - 5 320
AK311829.1 - 6 442
BC040107.1 - 5 330 5585294
BC052570.1 - 7 819
BC105808.1 - 6 446 6716437
BI600330.1 - 6 500
BX647867.1 - 5 323
DA821665.1 - 6 483
DA955978.1 - 7 543
DB472426.1 - 3 229
U51134.1 - 6 399

View Full Product Details