Gene Transcripts

Gene Symbol
Shc3
Entrez Gene ID
20418
Gene Name
src homology 2 domain-containing transforming protein C3
Gene Aliases
N-Shc, Rai, ShcC
Chromosome Location
Chr. 13: 51431041 - 51567084 on Build GRCm38
UniGene ID
Mm.131870
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009167.3 NP_033193.2 9 771
GenBank mRNA BC103611.1 - 9 771
BC103612.1 - 9 771 40046306
BC105644.1 - 9 771 40046301
BC105645.1 - 9 771 40046303
U46854.1 - 9 771

View Full Product Details