Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009383.2 | NP_033409.1 | 11, 12 | 1404 | |
GenBank mRNA | AK012085.1 | - | 11, 12 | 1048 | |
AK054473.1 | - | 4, 5 | 800 | ||
AK079363.1 | - | 1, 2 | 51 | ||
AK132677.1 | - | 12, 13 | 1572 | ||
BC010496.1 | - | 12, 13 | 1138 | 3497971 | |
BC068194.1 | - | 7, 8 | 680 | ||
U55861.1 | - | 11, 12 | 1180 |