Gene Transcripts

Gene Symbol
Phgr1
Entrez Gene ID
53906
Gene Name
proline/histidine/glycine-rich 1
Gene Aliases
1810007E14Rik, AA238765
Chromosome Location
Chr. 2: 118772769 - 118778164 on Build GRCm38
UniGene ID
Mm.36777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145644.1 NP_001139116.1 4 531
GenBank mRNA AK019007.1 - 4 527

View Full Product Details