Gene Transcripts

Gene Symbol
Tm4sf20
Entrez Gene ID
66261
Gene Name
transmembrane 4 L six family member 20
Gene Aliases
1810018L02Rik
Chromosome Location
Chr. 1: 82756649 - 82768456 on Build GRCm38
UniGene ID
Mm.46325
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025453.3 NP_079729.1 4 1243
GenBank mRNA AK007532.1 - 4 1245
AK017195.1 - 4 1244

View Full Product Details