Gene Transcripts
- Gene Symbol
- Srsf6
- Entrez Gene ID
- 67996
- Gene Name
- serine/arginine-rich splicing factor 6
- Gene Aliases
- 1210001E11Rik, AI314910, AW146126, Sfrs6
- Chromosome Location
- Chr. 2: 162931508 - 162937121 on Build GRCm38
- UniGene ID
- Mm.24042
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026499.4 |
NP_080775.3
|
3, 4 |
552 |
|
GenBank mRNA |
AK004831.1 |
- |
3, 4 |
533 |
|
AK012605.1 |
- |
3, 4 |
300 |
|
AK051608.1 |
- |
1, 2 |
705 |
|
AK133872.1 |
- |
3, 4 |
533 |
|
AK146293.1 |
- |
3, 4 |
530 |
|
AK159519.1 |
- |
3, 4 |
530 |
|
AK171709.1 |
- |
2, 3 |
1816 |
|
AK179065.1 |
- |
2, 3 |
181 |
|
AK179529.1 |
- |
1, 2 |
137 |
|
BC012039.1 |
- |
3, 4 |
539 |
2648111 |
View Full Product Details