Gene Transcripts
- Gene Symbol
- Vmn1r28
- Entrez Gene ID
- 171198
- Gene Name
- vomeronasal 1 receptor 28
- Gene Aliases
- V1rc25
- Chromosome Location
- Chr. 6: 58262294 - 58266082 on Build GRCm38
- UniGene ID
- Mm.222640
- Species
- Mus musculus
- Gene Symbol
- Ap2a1
- Entrez Gene ID
- 11771
- Gene Name
- adaptor-related protein complex 2, alpha 1 subunit
- Gene Aliases
- Adtaa
- Chromosome Location
- Chr. 7: 44900373 - 44929490 on Build GRCm38
- UniGene ID
- Mm.6877
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC114386.1 |
- |
|
663 |
|
View Full Product Details